Learn More
Cayman Chemical Δ2-Avermectin B1a, Purity: ≥95%, 5 mg
SDP
Catalog No. 502934166
Encompass_Preferred
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 0.23 and 0.038 ppm, respectively)
Catalog No. 50-293-4166 Supplier Cayman Chemical Supplier No. 251195 MG
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 0.23 and 0.038 ppm, respectively)