Learn More
Medchemexpress LLC (2-azabicyclo[2.1.1]hexan-1-yl)methanol hydrochloride | 2099148-27-1 | MFCD30833934 | 99.9% | 149.62 g·mol-1 | C6H12ClNO | 100 MG
SDP
Catalog No. 5000285270
Encompass_Preferred
(2-Azabicyclo[2.1.1]Hexan-1-yl)methanol hydrochloride is the hydrochloride salt of a bicyclic aminomethanol used as a synthetic building block and drug intermediate in medicinal chemistry. It is supplied as a solid for research use; molecular formula C6H12ClNO, molecular weight 149.62 g·mol-1, CAS 2099148-27-1. Follow SDS guidance for handling and storage.
  • High purity (COA indicates ~99.9%) for synthetic applications
  • Hydrochloride salt form eases handling and solubility control
  • Available in small pack sizes suitable for research-scale synthesis
  • Solid form with recommended sealed, cool storage to preserve stability
  • Documented analytical data and SDS available for compliance
Catalog No. 50-002-85270 Supplier Medchemexpress LLC Supplier No. HYW393807100MG
(2-Azabicyclo[2.1.1]Hexan-1-yl)methanol hydrochloride is the hydrochloride salt of a bicyclic aminomethanol used as a synthetic building block and drug intermediate in medicinal chemistry. It is supplied as a solid for research use; molecular formula C6H12ClNO, molecular weight 149.62 g·mol-1, CAS 2099148-27-1. Follow SDS guidance for handling and storage.
  • High purity (COA indicates ~99.9%) for synthetic applications
  • Hydrochloride salt form eases handling and solubility control
  • Available in small pack sizes suitable for research-scale synthesis
  • Solid form with recommended sealed, cool storage to preserve stability
  • Documented analytical data and SDS available for compliance