Learn More
Medchemexpress LLC (2-azabicyclo[2.1.1]hexan-1-yl)methanol hydrochloride | 2099148-27-1 | MFCD30833934 | 99.9% | 149.62 g/mol | C6H12ClNO | 250 MG
SDP
Catalog No. 5000285151
Encompass_Preferred
(2-Azabicyclo[2.1.1]Hexan-1-yl)methanol hydrochloride is a bicyclic amino alcohol supplied as a hydrochloride salt and used primarily as a chemical intermediate in medicinal chemistry and organic synthesis. It is supplied for research use only and should be handled following the safety data sheet and certificate of analysis.
  • High purity suitable for synthetic applications (99.9% by HPLC).
  • Available in small research pack sizes (e.g., 250 mg).
  • Stable when stored sealed at 4°C, away from moisture.
  • Provided with COA and SDS documentation for traceability.
  • Compatible with common organic solvents for low-temperature storage.
Catalog No. 50-002-85151 Supplier Medchemexpress LLC Supplier No. HYW393807250MG
(2-Azabicyclo[2.1.1]Hexan-1-yl)methanol hydrochloride is a bicyclic amino alcohol supplied as a hydrochloride salt and used primarily as a chemical intermediate in medicinal chemistry and organic synthesis. It is supplied for research use only and should be handled following the safety data sheet and certificate of analysis.
  • High purity suitable for synthetic applications (99.9% by HPLC).
  • Available in small research pack sizes (e.g., 250 mg).
  • Stable when stored sealed at 4°C, away from moisture.
  • Provided with COA and SDS documentation for traceability.
  • Compatible with common organic solvents for low-temperature storage.