Learn More
Medchemexpress LLC (2-azabicyclo[2.1.1]hexan-1-yl)methanol hydrochloride | 2099148-27-1 | MFCD30833934 | 99.9% | 149.62 g/mol | C6H12ClNO | 500 MG
SDP
Catalog No. 5000285037
Encompass_Preferred
(2-Azabicyclo[2.1.1]Hexan-1-yl)methanol hydrochloride is a chemical intermediate used in medicinal chemistry and small-scale synthesis of pharmacologically active compounds. Provided as the hydrochloride salt, the material is characterized by documented analytical data and storage recommendations to support research use.
  • High purity suitable for synthetic applications.
  • Available in multiple small-scale pack sizes for research and development.
  • Provided as a hydrochloride salt for improved handling and solubility.
  • Accompanied by certificate of analysis and safety data for quality and compliance.
  • Recommended storage: 4°C, sealed, away from moisture; solvent storage guidelines available.
  • Intended for use as a building block in drug intermediate synthesis.
Catalog No. 50-002-85037 Supplier Medchemexpress LLC Supplier No. HYW393807500MG
(2-Azabicyclo[2.1.1]Hexan-1-yl)methanol hydrochloride is a chemical intermediate used in medicinal chemistry and small-scale synthesis of pharmacologically active compounds. Provided as the hydrochloride salt, the material is characterized by documented analytical data and storage recommendations to support research use.
  • High purity suitable for synthetic applications.
  • Available in multiple small-scale pack sizes for research and development.
  • Provided as a hydrochloride salt for improved handling and solubility.
  • Accompanied by certificate of analysis and safety data for quality and compliance.
  • Recommended storage: 4°C, sealed, away from moisture; solvent storage guidelines available.
  • Intended for use as a building block in drug intermediate synthesis.