Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Azetidinone, 98%

Catalog No. AC432960010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432960010 1 g Glass bottle
AC432960050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432960010 Supplier Thermo Scientific Chemicals Supplier No. 432960010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 930-21-2 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-21-2
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00013328
InChI Key MNFORVFSTILPAW-UHFFFAOYSA-N
Synonym 2-azetidinone, beta-propiolactam, azetidinone, propiolactam, azetan-2-one, beta-lactam, 2-azetdinone, azetidine-2-one, acmc-209xab
PubChem CID 136721
ChEBI CHEBI:327119
IUPAC Name azetidin-2-one
SMILES C1CNC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 72.5°C to 76.5°C
Boiling Point 106°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 71.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Azetidinone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air a

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.