Learn More
Medchemexpress LLC 2-azido-2-deoxy-D-galactose | 68733-26-6 | MFCD09750682 | 98.0% | 205.17 | C6H11N3O5 | 10 MG
SDP
Catalog No. 5000300227
Encompass_Preferred
2-Azido-2-deoxy-D-galactose is an azido-functionalized galactose derivative supplied at 98.0% purity, used as a synthetic building block in glycobiology and bioorthogonal chemistry for the preparation of glycoconjugates and labeled glycans.
  • High purity (98.0%) suitable for research use.
  • Azide functional group enables bioorthogonal click chemistry.
  • Compatible with glycopeptide glycosylation and glycoconjugate synthesis.
  • Soluble in water (~25 mg/mL; ultrasonic assistance recommended).
  • Powder storage stability: -20°C (long term) and 4°C (short term).
  • Supplied in research-scale package sizes for laboratory applications.
Catalog No. 50-003-00227 Supplier Medchemexpress LLC Supplier No. HYW03991710MG
2-Azido-2-deoxy-D-galactose is an azido-functionalized galactose derivative supplied at 98.0% purity, used as a synthetic building block in glycobiology and bioorthogonal chemistry for the preparation of glycoconjugates and labeled glycans.
  • High purity (98.0%) suitable for research use.
  • Azide functional group enables bioorthogonal click chemistry.
  • Compatible with glycopeptide glycosylation and glycoconjugate synthesis.
  • Soluble in water (~25 mg/mL; ultrasonic assistance recommended).
  • Powder storage stability: -20°C (long term) and 4°C (short term).
  • Supplied in research-scale package sizes for laboratory applications.