Learn More
Medchemexpress LLC 2-azido-2-deoxy-D-galactose | 68733-26-6 | MFCD09750682 | 98.0% | 205.17 g/mol | C6H11N3O5 | 1 MG
SDP
Catalog No. 5000301259
Encompass_Preferred
2-Azido-2-deoxy-D-galactose is an azido-modified monosaccharide used in glycobiology research. The azide handle enables bioorthogonal chemistry for incorporation into glycopeptides and the synthesis of glycoconjugates. Supplied as a solid for research use only.
  • Azide functional group suitable for click chemistry and other bioorthogonal reactions.
  • Supports enzymatic or chemical glycosylation of peptides and glycans.
  • High purity for reliable analytical and synthetic applications.
  • Available in small-scale quantities for exploratory studies.
  • Supplied as a solid stable under recommended storage conditions.
Catalog No. 50-003-01259 Supplier Medchemexpress LLC Supplier No. HYW0399171MG
2-Azido-2-deoxy-D-galactose is an azido-modified monosaccharide used in glycobiology research. The azide handle enables bioorthogonal chemistry for incorporation into glycopeptides and the synthesis of glycoconjugates. Supplied as a solid for research use only.
  • Azide functional group suitable for click chemistry and other bioorthogonal reactions.
  • Supports enzymatic or chemical glycosylation of peptides and glycans.
  • High purity for reliable analytical and synthetic applications.
  • Available in small-scale quantities for exploratory studies.
  • Supplied as a solid stable under recommended storage conditions.