Learn More
Medchemexpress LLC 2-azido-2-deoxy-D-galactose | 68733-26-6 | MFCD09750682 | 98.0% | 205.17 g/mol | C6H11N3O5 | 5 MG
SDP
Catalog No. 5000299782
Encompass_Preferred
2-Azido-2-deoxy-D-galactose is an azido-functionalized galactose monosaccharide used in glycobiology research. The azide handle enables bioorthogonal labeling and conjugation, and the compound serves as a building block for enzymatic or chemical glycosylation to synthesize labeled glycoconjugates.
  • Azido-functionalized monosaccharide for glycobiology applications.
  • Enables bioorthogonal conjugation such as copper-catalyzed or strain-promoted click chemistry.
  • Can be glycosylated by glycopeptides to build glycoconjugates.
  • High chemical purity (98.0%).
  • Water-soluble (≈25 mg/mL; may require ultrasonic agitation).
  • Stable as a powder under cold storage; in-solution storage requires low temperatures.
Catalog No. 50-002-99782 Supplier Medchemexpress LLC Supplier No. HYW0399175MG
2-Azido-2-deoxy-D-galactose is an azido-functionalized galactose monosaccharide used in glycobiology research. The azide handle enables bioorthogonal labeling and conjugation, and the compound serves as a building block for enzymatic or chemical glycosylation to synthesize labeled glycoconjugates.
  • Azido-functionalized monosaccharide for glycobiology applications.
  • Enables bioorthogonal conjugation such as copper-catalyzed or strain-promoted click chemistry.
  • Can be glycosylated by glycopeptides to build glycoconjugates.
  • High chemical purity (98.0%).
  • Water-soluble (≈25 mg/mL; may require ultrasonic agitation).
  • Stable as a powder under cold storage; in-solution storage requires low temperatures.