Learn More
Medchemexpress LLC 2-azido-2-deoxy-D-galactose | 68733-26-6 | MFCD09750682 | 98.0% | 205.17 g·mol⁻¹ | C6H11N3O5 | 50 MG
SDP
Catalog No. 5000301254
Encompass_Preferred
2-Azido-2-deoxy-D-galactose (98%) is an azido-functionalized galactose derivative provided as a white to off-white solid for laboratory research. It is used as a reactive sugar building block for incorporation into glycans, glycopeptides, and glycoconjugates, and is supported by analytical documentation for quality assurance.
  • High purity (98.0%).
  • Azido functional group enables bioorthogonal chemistry.
  • Compatible with enzymatic and chemical glycosylation.
  • Supplied as a stable, easy-to-handle solid.
  • COA, SDS, and spectral reports available for verification.
Catalog No. 50-003-01254 Supplier Medchemexpress LLC Supplier No. HYW03991750MG
2-Azido-2-deoxy-D-galactose (98%) is an azido-functionalized galactose derivative provided as a white to off-white solid for laboratory research. It is used as a reactive sugar building block for incorporation into glycans, glycopeptides, and glycoconjugates, and is supported by analytical documentation for quality assurance.
  • High purity (98.0%).
  • Azido functional group enables bioorthogonal chemistry.
  • Compatible with enzymatic and chemical glycosylation.
  • Supplied as a stable, easy-to-handle solid.
  • COA, SDS, and spectral reports available for verification.