Learn More
Medchemexpress LLC 2'-Azido-2'-deoxycytidine | 51034-68-5 | C9H12N6O4 | 100 MG
SDP
Catalog No. 5000373387
Encompass_Preferred
2'-Azido-2'-deoxycytidine is a purine nucleoside analogue with broad antitumor activity, especially against indolent lymphoid malignancies, acting by inhibiting DNA synthesis and inducing apoptosis. It serves as a click chemistry reagent, possessing an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. Diphosphorylation is required for its function as a nucleotide reductase inhibitor.
  • Purine nucleoside analogue with antitumor activity
  • Inhibits DNA synthesis and induces apoptosis
  • Click chemistry reagent for CuAAc and SPAAC reactions
  • Requires diphosphorylation to inhibit nucleotide reductase
  • Useful in oligonucleotide and nucleoside analog research
  • Applicable in cancer studies, including leukemia
  • Relevant for cell cycle and DNA damage investigations
Catalog No. 50-003-73387 Supplier Medchemexpress LLC Supplier No. HY152448100MG
2'-Azido-2'-deoxycytidine is a purine nucleoside analogue with broad antitumor activity, especially against indolent lymphoid malignancies, acting by inhibiting DNA synthesis and inducing apoptosis. It serves as a click chemistry reagent, possessing an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. Diphosphorylation is required for its function as a nucleotide reductase inhibitor.
  • Purine nucleoside analogue with antitumor activity
  • Inhibits DNA synthesis and induces apoptosis
  • Click chemistry reagent for CuAAc and SPAAC reactions
  • Requires diphosphorylation to inhibit nucleotide reductase
  • Useful in oligonucleotide and nucleoside analog research
  • Applicable in cancer studies, including leukemia
  • Relevant for cell cycle and DNA damage investigations