Learn More
Medchemexpress LLC 2-((Azido-PEG8-carbamoyl)methoxy)acetic acid | 846549-37-9 | 98.6% | C22H42N4O12 | 1 G
SDP
Catalog No. 5000356990
Encompass_Preferred
2-((Azido-PEG8-carbamoyl)methoxy)acetic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules. It also participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Features an Azide group for CuAAc reactions
  • Participates in SPAAC reactions
  • Relevant to click chemistry and PROTAC synthesis
  • For research use only
Catalog No. 50-003-56990 Supplier Medchemexpress LLC Supplier No. HY1404591G
2-((Azido-PEG8-carbamoyl)methoxy)acetic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules. It also participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Features an Azide group for CuAAc reactions
  • Participates in SPAAC reactions
  • Relevant to click chemistry and PROTAC synthesis
  • For research use only