Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Azidoethyl 2-bromoisobutyrate

Catalog No. 7920551G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-205-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-205-51G Supplier Sigma Aldrich Supplier No. 7920551G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bifunctional initiator with a bromoisobutyryl moiety for atom transfer radical polymerization (ATRP) and an azide moiety that can be used in Cu-mediated ligation (click chemistry) for biomaterials, carbon nanotubes and graphene sheets.

Specifications

CAS 1120364-53-5
Density 1.437 g/mL (at 25°C)
Molecular Formula C6H10BrN3O2
Refractive Index n20/D 1.482
Linear Formula C6H10BrN3O2
Quantity 1 g
Synonym 2 -Azidoethyl 2-bromoisobutyrate; 2-Azidoethyl 2-bromo-2-methylpropanoate; 2-Azidoethyl-2-bromoisobutyrate; 2-Bromo-2-methyl-propanoic acid 2-azidoethyl ester; Azido-ATRP initiator; Azido-Bib
Molecular Weight (g/mol) 236.07
Percent Purity 97%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.