Learn More
Medchemexpress LLC 2-azidoethyl β-D-lactoside | 230286-11-0 | MFCD19981033 | 95.0% | 411.36 g/mol | C14H25N3O11 | 10 MG
SDP
Catalog No. 5000286502
Encompass_Preferred
2-Azidoethyl β-D-lactoside is an azide-functionalized β-D-lactoside reagent designed for click-chemistry conjugation. It reacts with alkyne-containing molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC), enabling efficient bioconjugation in glycochemistry and chemical biology workflows.
Catalog No. 50-002-86502 Supplier Medchemexpress LLC Supplier No. HY15179310MG
2-Azidoethyl β-D-lactoside is an azide-functionalized β-D-lactoside reagent designed for click-chemistry conjugation. It reacts with alkyne-containing molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC), enabling efficient bioconjugation in glycochemistry and chemical biology workflows.