Learn More
Medchemexpress LLC 2-Azidoethyl β-D-lactoside | 50 MG
SDP
Catalog No. 5000374936
Encompass_Preferred
2-Azidoethyl β-D-lactoside (beta-Lac-EO-N3) is a click chemistry reagent suitable for various research fields. Click chemistry offers beneficial characteristics such as high yield, high specificity, and simplicity, making it ideal for binding between nucleic acids, lipids, proteins, and other molecules.
  • Contains an azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups
  • Exhibits high yield in reactions
  • Offers high specificity
  • Provides simplicity in experimental procedures
Catalog No. 50-003-74936 Supplier Medchemexpress LLC Supplier No. HY15179350MG
2-Azidoethyl β-D-lactoside (beta-Lac-EO-N3) is a click chemistry reagent suitable for various research fields. Click chemistry offers beneficial characteristics such as high yield, high specificity, and simplicity, making it ideal for binding between nucleic acids, lipids, proteins, and other molecules.
  • Contains an azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups
  • Exhibits high yield in reactions
  • Offers high specificity
  • Provides simplicity in experimental procedures