Learn More
Chemglass Life Sciences Coupling, 2" Beaded Pipe, "A" 3-1/8" X "B" 1-5/16", 60 in-lbs MAX Torque, 7/16" Hex Nut Size
SDP
Catalog No. CG196867
Encompass_Preferred
Coupling, 2" Beaded Pipe, "A" 3-1/8" X "B" 1-5/16", 60 in-lbs MAX Torque, 7/16" Hex Nut Size. Complete coupling assembly consists of a 300 series stainless steel outer band with T-bolt, a Viton liner, and a PTFE sealing ring. Couplings allow for a maximum deflection of 4¿. PLEASE NOTE: TIGHTEN ALL COUPLINGS TO THE PROPER TORQUE SETTING.
Catalog No. CG196867 Supplier Chemglass Life Sciences Supplier No. CG196867
Coupling, 2" Beaded Pipe, "A" 3-1/8" X "B" 1-5/16", 60 in-lbs MAX Torque, 7/16" Hex Nut Size. Complete coupling assembly consists of a 300 series stainless steel outer band with T-bolt, a Viton liner, and a PTFE sealing ring. Couplings allow for a maximum deflection of 4¿. PLEASE NOTE: TIGHTEN ALL COUPLINGS TO THE PROPER TORQUE SETTING.