Learn More
Medchemexpress LLC 2-benzamidoacetic acid-d5 | 53518-98-2 | 99.7% | 184.20 g/mol | C9H4D5NO3 | 1 MG
SDP
Catalog No. 5000301831
Encompass_Preferred
Hippuric acid-d5 is a deuterium-labeled analogue of hippuric acid used as a stable isotope internal standard for quantitative mass spectrometry and metabolic studies. It serves as a reference standard for analytical method development, validation, and tracer experiments. Molecular formula: C9H4D5NO3; CAS: 53518-98-2.
  • Deuterium-labeled internal standard for LC-MS/MS.
  • High isotopic enrichment for accurate quantitation.
  • Suitable for method development and validation.
  • Applicable to metabolic tracing and biomarker studies.
  • Provided in small quantities for analytical use.
Catalog No. 50-003-01831 Supplier Medchemexpress LLC Supplier No. HYW016562S1MG
Hippuric acid-d5 is a deuterium-labeled analogue of hippuric acid used as a stable isotope internal standard for quantitative mass spectrometry and metabolic studies. It serves as a reference standard for analytical method development, validation, and tracer experiments. Molecular formula: C9H4D5NO3; CAS: 53518-98-2.
  • Deuterium-labeled internal standard for LC-MS/MS.
  • High isotopic enrichment for accurate quantitation.
  • Suitable for method development and validation.
  • Applicable to metabolic tracing and biomarker studies.
  • Provided in small quantities for analytical use.