Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Benzenesulfonylmethyl-1H-benzoimidazole

Catalog No. CDS011700 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS011700 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS011700 Supplier Sigma Aldrich Supplier No. CDS011700100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans-5-decene has been used to characterize the effect of cyclization and size on the stability of criegee intermediates formed in ozonolysis
  • It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts

Specifications

CAS 21094-70-2
Molecular Formula C14H12N2O2S
Linear Formula C14H12N2O2S
Quantity 5 g
MDL Number MFCD02044407
Molecular Weight (g/mol) 272.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.