Learn More
Medchemexpress LLC 2-((Benzo[d]thiazol-2-ylmethyl)thio)-4,5,6,7-tetrahydro-8H-cyclopenta[d]triazolo[1,5-a]pyrimidin-8 | 3112403-98-9 | 99.9% | 355.44 | 25 MG
SDP
Catalog No. 5000393931
Encompass_Preferred
WS-917 is a potent and orally active ABCB1 modulator that enhances ABCB1 ATPase activity. It significantly enhances the antitumor effect of paclitaxel when combined with an anti-PD-1 antibody, and also promotes CD8+ T-cell activation. This product is for research use only and has not been fully validated for medical applications.
  • Potent and orally active ABCB1 modulator
  • Enhances ABCB1 ATPase activity
  • Significantly enhances antitumor effect of paclitaxel combined with anti-PD-1 antibody
  • Promotes CD8+ T-cell activation
Catalog No. 50-003-93931 Supplier Medchemexpress LLC Supplier No. 25MGHY168528
WS-917 is a potent and orally active ABCB1 modulator that enhances ABCB1 ATPase activity. It significantly enhances the antitumor effect of paclitaxel when combined with an anti-PD-1 antibody, and also promotes CD8+ T-cell activation. This product is for research use only and has not been fully validated for medical applications.
  • Potent and orally active ABCB1 modulator
  • Enhances ABCB1 ATPase activity
  • Significantly enhances antitumor effect of paclitaxel combined with anti-PD-1 antibody
  • Promotes CD8+ T-cell activation