Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Benzofurancarboxylic acid,5-(1-piperazinyl)-,ethyl ester | 163521-20-8 | 99.9% | C15H18N2O3 | 25 G
SDP

Catalog No. 5000358654
Encompass_Preferred

Ethyl 5-piperazin-1-ylbenzofuran-2-carboxylate is a white to yellow solid, tested and verified to comply with specifications.

  • Appearance: White to yellow solid
  • Purity (HPLC): 99.88%
  • Molecular formula: C15H18N2O3
  • Molecular weight: 274.32
  • Storage: 4°C, protected from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month, protected from light

Catalog No. 50-003-58654 Supplier Medchemexpress LLC Supplier No. 25GHY77058
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 5-piperazin-1-ylbenzofuran-2-carboxylate is a white to yellow solid, tested and verified to comply with specifications.

  • Appearance: White to yellow solid
  • Purity (HPLC): 99.88%
  • Molecular formula: C15H18N2O3
  • Molecular weight: 274.32
  • Storage: 4°C, protected from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.