Learn More
Medchemexpress LLC 2-benzofurancarboxylic acid, 5-(1-piperazinyl)-,ethyl ester | 163521-20-8 | 99.9% | C15H18N2O3 | 5 G
SDP
Catalog No. 5000433536
Encompass_Preferred
Ethyl 5-piperazin-1-ylbenzofuran-2-carboxylate (CAS 163521-20-8) is a chemical compound supplied as a white to yellow solid. It has a molecular formula of C15H18N2O3 and a molecular weight of 274.32. This product is suitable for research use.
  • High purity of 99.88% (HPLC)
  • White to yellow solid appearance
  • Storage at 4°C, protected from light
  • In solvent storage conditions: -80°C for 6 months; -20°C for 1 month (protect from light)
  • For research use only
Catalog No. 50-004-33536 Supplier Medchemexpress LLC Supplier No. HY770585G
Add to Cart
Add to Cart
Ethyl 5-piperazin-1-ylbenzofuran-2-carboxylate (CAS 163521-20-8) is a chemical compound supplied as a white to yellow solid. It has a molecular formula of C15H18N2O3 and a molecular weight of 274.32. This product is suitable for research use.
  • High purity of 99.88% (HPLC)
  • White to yellow solid appearance
  • Storage at 4°C, protected from light
  • In solvent storage conditions: -80°C for 6 months; -20°C for 1 month (protect from light)
  • For research use only