Learn More
Apexbio Technology LLC 2-BENZOXAZOLINONE-100MG
SDP
Catalog No. 5000569419
Encompass_Preferred
2-Benzoxazolinone (CAS No 59-49-4) is a heterocyclic organic compound widely studied in the field of biomedical research It originates from a benzoxazole structure known for its role in modulating various biological pathways including enzyme inhibition and oxidative stress responses Its mechanism of action often involves interference with specific cellular targets though detailed pathways can vary based on the research context In vitro it demonstrates activity usually within the nanomolar to low micromolar range Common applications include assays involving cell models to understand its biochemical interactions and animal studies to explore therapeutic potentials and toxicology It is also utilized in drug screening efforts as a potential lead compound Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69419 Supplier Apexbio Technology LLC Supplier No. C7114100
Add to Cart
Add to Cart
2-Benzoxazolinone (CAS No 59-49-4) is a heterocyclic organic compound widely studied in the field of biomedical research It originates from a benzoxazole structure known for its role in modulating various biological pathways including enzyme inhibition and oxidative stress responses Its mechanism of action often involves interference with specific cellular targets though detailed pathways can vary based on the research context In vitro it demonstrates activity usually within the nanomolar to low micromolar range Common applications include assays involving cell models to understand its biochemical interactions and animal studies to explore therapeutic potentials and toxicology It is also utilized in drug screening efforts as a potential lead compound Experimental concentrations typically depend on the specific research design