Learn More
Medchemexpress LLC HY-W015818 100mg Medchemexpress, 2-Benzoxazolinone CAS:59-49-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW015818100MG
Encompass_Preferred
Medchemexpress, HY-W015818 100mg 2-Benzoxazolinone CAS:59-49-4 2-Benzoxazolinone is an anti-leishmanial agent with an LC 50 of 40 μg/mL against L. donovani [1] . A building block in chemical synthesis. 1,3-Benzoxazol-2(3H)-one derivatives have antimicrobial activity against a selection of Gram-positive, Gram-negative bacteria and yeasts [3] . Derivatives as anti-quorum sensing agent [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4609