Learn More
Medchemexpress LLC 2-benzoxazolinone | 59-49-4 | MFCD00005716 | 99.9% | 135.12 g/mol | C7H5NO2 | 1 ML
SDP
Catalog No. 5000300770
Encompass_Preferred
2-Benzoxazolinone (C7H5NO2; MW 135.12 g/mol) is a small-molecule heterocycle supplied as a 10 mM solution in DMSO for use in biological screening and synthetic chemistry. It has reported antiparasitic activity and is commonly used as a building block in organic synthesis; typical purity is 99.87%.
  • Pre-made 10 mM solution in DMSO for immediate use
  • High purity (≈99.9%) suitable for research applications
  • Well-characterized chemical: C7H5NO2, 135.12 g/mol
  • Reported bioactivity including antiparasitic effects
  • Convenient for screening, assay development, and synthetic work
Catalog No. 50-003-00770 Supplier Medchemexpress LLC Supplier No. HYW01581810MM/1ML
2-Benzoxazolinone (C7H5NO2; MW 135.12 g/mol) is a small-molecule heterocycle supplied as a 10 mM solution in DMSO for use in biological screening and synthetic chemistry. It has reported antiparasitic activity and is commonly used as a building block in organic synthesis; typical purity is 99.87%.
  • Pre-made 10 mM solution in DMSO for immediate use
  • High purity (≈99.9%) suitable for research applications
  • Well-characterized chemical: C7H5NO2, 135.12 g/mol
  • Reported bioactivity including antiparasitic effects
  • Convenient for screening, assay development, and synthetic work