Learn More
Medchemexpress LLC 2-benzoxazolinone | 59-49-4 | MFCD00005716 | 99.9% | 135.12 g/mol | C7H5NO2 | 100 G
SDP
Catalog No. 5000300769
Encompass_Preferred
2-Benzoxazolinone is a small-molecule research chemical and synthetic building block (CAS 59-49-4) with reported anti-leishmanial activity. It is supplied in high purity and offered as both a solid and a ready-to-use DMSO solution for analytical and synthetic applications.
  • High purity (99.87%) suitable for research use.
  • Available as solid (50 g, 100 g, 500 g) and as 10 mM solution in DMSO.
  • Useful building block for organic synthesis.
  • Reported anti-leishmanial activity (LC50 40 μg/mL).
  • Well characterized: formula C7H5NO2, molecular weight 135.12 g/mol.
Catalog No. 50-003-00769 Supplier Medchemexpress LLC Supplier No. HYW015818100G
2-Benzoxazolinone is a small-molecule research chemical and synthetic building block (CAS 59-49-4) with reported anti-leishmanial activity. It is supplied in high purity and offered as both a solid and a ready-to-use DMSO solution for analytical and synthetic applications.
  • High purity (99.87%) suitable for research use.
  • Available as solid (50 g, 100 g, 500 g) and as 10 mM solution in DMSO.
  • Useful building block for organic synthesis.
  • Reported anti-leishmanial activity (LC50 40 μg/mL).
  • Well characterized: formula C7H5NO2, molecular weight 135.12 g/mol.