Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-benzoxazolinone | 59-49-4 | MFCD00005716 | 99.9% | 135.12 g/mol | C7H5NO2 | 50 G
SDP

Catalog No. 5000300330
Encompass_Preferred

2-Benzoxazolinone (CAS 59-49-4) is a small-molecule reagent used as a building block in organic synthesis and reported to have anti-leishmanial activity. It is supplied as a high-purity solid for research and analytical applications, with documented solubility and storage recommendations.

  • High purity (99.9%).
  • Molecular formula C7H5NO2; molecular weight 135.12 g/mol.
  • Soluble in DMSO (≈100 mg/mL) and sparingly soluble in water with sonication.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable as a synthetic building block and for biological screening.

Catalog No. 50-003-00330 Supplier Medchemexpress LLC Supplier No. HYW01581850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Benzoxazolinone (CAS 59-49-4) is a small-molecule reagent used as a building block in organic synthesis and reported to have anti-leishmanial activity. It is supplied as a high-purity solid for research and analytical applications, with documented solubility and storage recommendations.

  • High purity (99.9%).
  • Molecular formula C7H5NO2; molecular weight 135.12 g/mol.
  • Soluble in DMSO (≈100 mg/mL) and sparingly soluble in water with sonication.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable as a synthetic building block and for biological screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.