Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-benzoxazolinone | 59-49-4 | MFCD00005716 | 99.9% | 135.12 g/mol | C7H5NO2 | 500 G
SDP

Catalog No. 5000300696
Encompass_Preferred

2-Benzoxazolinone is a small-molecule research reagent (C7H5NO2) used as a synthetic building block and biological probe. It has reported anti-leishmanial activity and its derivatives exhibit antimicrobial and anti-quorum sensing properties. The compound is supplied as a stable powder with supporting documentation (COA, SDS, datasheet) suitable for chemical synthesis and biological research.

  • CAS number: 59-49-4.
  • Purity: 99.87%.
  • Molecular weight: 135.12 g/mol.
  • Molecular formula: C7H5NO2.
  • Solubility: soluble in DMSO (~100 mg/mL); limited solubility in water (~3.85 mg/mL).
  • Storage: powder stable at -20°C; stock solutions stable at -80°C.
  • Applications: synthetic building block and biological research reagent.

Catalog No. 50-003-00696 Supplier Medchemexpress LLC Supplier No. HYW015818500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Benzoxazolinone is a small-molecule research reagent (C7H5NO2) used as a synthetic building block and biological probe. It has reported anti-leishmanial activity and its derivatives exhibit antimicrobial and anti-quorum sensing properties. The compound is supplied as a stable powder with supporting documentation (COA, SDS, datasheet) suitable for chemical synthesis and biological research.

  • CAS number: 59-49-4.
  • Purity: 99.87%.
  • Molecular weight: 135.12 g/mol.
  • Molecular formula: C7H5NO2.
  • Solubility: soluble in DMSO (~100 mg/mL); limited solubility in water (~3.85 mg/mL).
  • Storage: powder stable at -20°C; stock solutions stable at -80°C.
  • Applications: synthetic building block and biological research reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.