Learn More
Selleck Chemical LLC 2-Benzoxazolinone
SDP
Catalog No. 5000004008
Encompass_Preferred
2-Benzoxazolinone (BOA 2-Hydroxybenzoxazole) a well-known allelochemical with strong phytotoxicity is a potential herbicidal candidate and probably functions via interfering with auxins or disrupting membrane integrity
Catalog No. 50-000-04008 Supplier Selleck Chemical LLC Supplier No. S557525MG
2-Benzoxazolinone (BOA 2-Hydroxybenzoxazole) a well-known allelochemical with strong phytotoxicity is a potential herbicidal candidate and probably functions via interfering with auxins or disrupting membrane integrity