Learn More
Medchemexpress LLC 2-Benzoylpyridine | 91-02-1 | MFCD00006300 | 183.21 g/mol | C12H9NO | 500 G

Supplier: Medchemexpress LLC HYW042192500G
2-Benzoylpyridine is a small-molecule biochemical reagent intended for research and analytical applications. It is provided for laboratory use as a reagent or standard; consult supplier documentation and safety data sheets for handling and storage information.
- Used as a reagent or analytical standard in research applications.
- Molecular formula C12H9NO, molecular weight 183.21 g/mol.
- CAS number 91-02-1 for unambiguous substance identification.
- Available in multiple small-package sizes per manufacturer; confirm quantity with supplier before purchase.
- For research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.