Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-benzoylpyridine | 91-02-1 | MFCD00006300 | >99.0% | 183.21 g/mol | C12H9NO | 50 G
SDP

Catalog No. 5000209262
Encompass_Preferred

2-benzoylpyridine is an organic compound used as a biochemical reagent and analytical standard. It has formula C12H9NO and a molecular weight of 183.21 g/mol. Common applications include use as a reagent in synthesis and as an analytical reference standard.

  • Chemical name: 2-benzoylpyridine
  • Use: biochemical reagent and analytical standard
  • CAS number: 91-02-1
  • Molecular formula: C12H9NO
  • Molecular weight: 183.21 g/mol
  • Typical purity: >99% (supplier-reported)

Catalog No. 50-002-09262 Supplier Medchemexpress LLC Supplier No. HYW04219250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-benzoylpyridine is an organic compound used as a biochemical reagent and analytical standard. It has formula C12H9NO and a molecular weight of 183.21 g/mol. Common applications include use as a reagent in synthesis and as an analytical reference standard.

  • Chemical name: 2-benzoylpyridine
  • Use: biochemical reagent and analytical standard
  • CAS number: 91-02-1
  • Molecular formula: C12H9NO
  • Molecular weight: 183.21 g/mol
  • Typical purity: >99% (supplier-reported)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.