Learn More
Medchemexpress LLC 2-benzoylpyridine | 91-02-1 | MFCD00006300 | 99.9% | 183.21 g/mol | C12H9NO | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0421921KG
Encompass_Preferred
2-Benzoylpyridine is a small-molecule biochemical reagent supplied for research use. It is used as an analytical standard, a synthetic intermediate, and in organic synthesis and analytical applications.
  • Used as an analytical standard and reference material.
  • Serves as a synthetic intermediate in organic synthesis.
  • Has CAS number 91-02-1, formula C12H9NO, and molecular weight 183.21 g/mol.
  • Available in small research sizes; larger quantities available by bulk inquiry.
  • Purity reported by third-party distributors as 99.92%; manufacturer datasheet does not list a numerical purity.
Catalog No. 50-002-09225