Learn More
Medchemexpress LLC 2-Benzoylpyridine | 91-02-1 | MFCD00006300 | 99.9% | 183.21 g/mol | C12H9NO | 25 G

Supplier: Medchemexpress LLC 25GHYW042192
2-Benzoylpyridine is a high-purity biochemical reagent used in organic synthesis and biochemical assays. It has molecular formula C12H9NO, a molecular weight of 183.21 g/mol, and is supplied as a 25 g package with reported purity of 99.9%.
- High purity of 99.9%.
- Used as a reagent in organic synthesis and biochemical assays.
- Molecular formula C12H9NO; molecular weight 183.21 g/mol.
- Available as a 25 g package for laboratory use.
- Supplied with product datasheet and analytical information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.