Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyl-4-chlorophenol 97.0+%, TCI America™
SDP

Catalog No. B081225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B081225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B081225G Supplier TCI America Supplier No. B081225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-32-1
Molecular Formula C13H11ClO
Molecular Weight (g/mol) 218.68
MDL Number MFCD00020140
InChI Key NCKMMSIFQUPKCK-UHFFFAOYSA-N
Synonym clorophene, chlorophene, clorofene, o-benzyl-p-chlorophenol, neosabenyl, septiphene, benzylchlorophenol, bio-clave, 4-chloro-2-benzylphenol, santophen 1
PubChem CID 8425
IUPAC Name 2-benzyl-4-chlorophenol
SMILES OC1=CC=C(Cl)C=C1CC1=CC=CC=C1

Specifications

Melting Point 47°C
Boiling Point 162°C
Quantity 25 g
UN Number 3077
Formula Weight 218.68
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyl-4-chlorophenol
EINECSNumber (4)-0098
RTECSNumber GO7175000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.