Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(benzylamino)acetic acid | 17136-36-6 | MFCD00190772 | 98.5% | 165.19 g/mol | C9H11NO2 | 25 G
SDP

Catalog No. 5000303425
Encompass_Preferred

2-(Benzylamino)acetic acid is a glycine-derived amino acid for research use supplied as a white to off-white solid. The product is characterized by its chemical identity (CAS 17136-36-6) and is provided with supporting documentation for laboratory handling and analysis.

  • Reported purity 98.46%.
  • Chemical formula C9H11NO2; molecular weight 165.19 g/mol.
  • Available in common pack sizes (25 g, 100 g).
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Storage recommendations provided for powder and solvent conditions.
  • White to off-white solid appearance.

Catalog No. 50-003-03425 Supplier Medchemexpress LLC Supplier No. 25GHYW015359
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Benzylamino)acetic acid is a glycine-derived amino acid for research use supplied as a white to off-white solid. The product is characterized by its chemical identity (CAS 17136-36-6) and is provided with supporting documentation for laboratory handling and analysis.

  • Reported purity 98.46%.
  • Chemical formula C9H11NO2; molecular weight 165.19 g/mol.
  • Available in common pack sizes (25 g, 100 g).
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Storage recommendations provided for powder and solvent conditions.
  • White to off-white solid appearance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.