Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzylaminopyridine 98.0+%, TCI America™
SDP

Catalog No. B120925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B120925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B120925G Supplier TCI America Supplier No. B120925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6935-27-9
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00006251
InChI Key WYHXNQXDQQMTQI-UHFFFAOYSA-N
Synonym 2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g
PubChem CID 23362
IUPAC Name N-benzylpyridin-2-amine
SMILES C1=CC=C(C=C1)CNC2=CC=CC=N2

Specifications

Melting Point 95°C
Boiling Point 182°C
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylaminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.