Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-benzylideneheptanal | 122-40-7 | MFCD00006988 | 99.6% | 202.29 g/mol | C14H18O | 1 ML
SDP

Catalog No. 5000302722
Encompass_Preferred

2-Benzylideneheptanal (α-amylcinnamaldehyde, CAS 122-40-7) is an aromatic aldehyde supplied as a ready-to-use 10 mM solution in DMSO for laboratory research. It has formula C14H18O, molecular weight 202.29 g/mol, and a reported purity of 99.56%. The solution is intended for in vitro biochemical or ligand screening and is provided with accompanying analytical documentation for handling and quality verification.

  • Ready-to-use 10 mM solution in DMSO, 1 mL vial.
  • High reported purity (99.6%).
  • Chemical identity: 2-benzylideneheptanal (CAS 122-40-7).
  • Molecular weight 202.29 g/mol; formula C14H18O.
  • Supplied with SDS, COA, and handling instructions.
  • Suitable for in vitro biochemical assays and ligand screening.

Catalog No. 50-003-02722 Supplier Medchemexpress LLC Supplier No. HYW01478010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Benzylideneheptanal (α-amylcinnamaldehyde, CAS 122-40-7) is an aromatic aldehyde supplied as a ready-to-use 10 mM solution in DMSO for laboratory research. It has formula C14H18O, molecular weight 202.29 g/mol, and a reported purity of 99.56%. The solution is intended for in vitro biochemical or ligand screening and is provided with accompanying analytical documentation for handling and quality verification.

  • Ready-to-use 10 mM solution in DMSO, 1 mL vial.
  • High reported purity (99.6%).
  • Chemical identity: 2-benzylideneheptanal (CAS 122-40-7).
  • Molecular weight 202.29 g/mol; formula C14H18O.
  • Supplied with SDS, COA, and handling instructions.
  • Suitable for in vitro biochemical assays and ligand screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.