Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Benzyloxy-5-methylphenylboronic acid pinacol ester

Catalog No. SYX001961G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX001961G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX001961G Supplier Sigma Aldrich Supplier No. SYX001961G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-iodononane has been used in the synthesis of:
  • Sch ii, a cerebroside that induces fruiting body formation in the basidiomycete schizophyllum commune
  • 2,6-dimethyl-4-nonylpyridine
  • 2-tridecanone

Specifications

CAS 1204580-85-7
Molecular Formula C20H25BO3
Linear Formula C20H25BO3
MDL Number MFCD13152011
Quantity 25 g
Molecular Weight (g/mol) 324.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.