Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxybenzeneboronic acid, 96%

Catalog No. AAL2010006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2010006 5 g
AAL2010003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2010006 Supplier Thermo Scientific Chemicals Supplier No. L2010006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 190661-29-1 | C13H13BO3 | 228.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190661-29-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.054
MDL Number MFCD01632206
InChI Key MCAIDINWZOCYQK-UHFFFAOYSA-N
Synonym 2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl
PubChem CID 2773253
IUPAC Name (2-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O

Specifications

Melting Point 96°C to 99°C
Quantity 5 g
Beilstein 7813704
Formula Weight 228.06
Percent Purity 96%
Chemical Name or Material 2-Benzyloxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.