Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxyphenylacetic acid, 98+%

Catalog No. AAA1523703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1523703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1523703 Supplier Thermo Scientific Chemicals Supplier No. A1523703

missing translation for 'validMainframeMsg'

CAS: 22047-88-7 | C15H13O3 | 241.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22047-88-7
Molecular Formula C15H13O3
Molecular Weight (g/mol) 241.27
MDL Number MFCD00016822
InChI Key BGTPJDGURCMYML-AWEZNQCLSA-M
Synonym 2-benzyloxyphenylacetic acid, o-benzyloxyphenylacetic acid, 2-2-benzyloxy phenyl acetic acid, 2-benzyloxy phenyl acetic acid, 2-benzyloxyphenyl acetic acid, benzeneacetic acid,2-phenylmethoxy, 2-2-phenylmethoxy phenyl acetic acid, acmc-20ao3j
PubChem CID 519916
SMILES [O-]C(=O)[C@@H](OCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C to 98°C
Quantity 1 g
Beilstein 2653977
IUPAC Name 2-(2-phenylmethoxyphenyl)acetic acid
Formula Weight 242.28
Percent Purity ≥98%
Chemical Name or Material 2-Benzyloxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.