Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B45905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4590-5G 5 g
B45901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4590-5G Supplier TCI America Supplier No. B45905G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 190661-29-1
Molecular Formula C13H13BO3
MDL Number MFCD01632206
InChI Key MCAIDINWZOCYQK-UHFFFAOYSA-N
PubChem CID 2773253

Specifications

Melting Point 110°C
Color White
Quantity 5 g
Synonym 2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid
SMILES OB(O)C1=CC=CC=C1OCC1=CC=CC=C1
IUPAC Name [2-(benzyloxy)phenyl]boronic acid
Molecular Weight (g/mol) 228.05
Formula Weight 228.05
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.