Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Benzyloxyphenylmagnesium Bromide Solution

Catalog No. 67301350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-301-350ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 67-301-350ML Supplier Sigma Aldrich Supplier No. 67301350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Benzyloxyphenylmagnesium bromide can be used:As a substrate in the iron-catalyzed coupling reaction of aryl Grignard reagents with unprotected bromophenols.As a substrate in the cross-coupling of Grignard reagents using diaryl titanates and ferric chloride.In one of the key synthetic steps for the total synthesis of an antibiotic caboxamycin.

Specifications

CAS 103765-03-3
Melting Point 258°C to 262°C
Molecular Formula C4H11ClN2O
Linear Formula CH3CH2CH(NH2)CONH2 -+ HCl
MDL Number MFCD09265126
Quantity 1 g
Synonym (R)-2-Aminobutyramide hydrochloride
Molecular Weight (g/mol) 138.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.