Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzylphenol 97.0+%, TCI America™
SDP

Catalog No. B0111500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0111500G 500 g
B011125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0111500G Supplier TCI America Supplier No. B0111500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28994-41-4
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00002242
InChI Key CDMGNVWZXRKJNS-UHFFFAOYSA-N
Synonym 2-hydroxydiphenylmethane, o-benzylphenol, phenol, 2-phenylmethyl, benzylphenol, 2-benzyl-phenol, 2-phenylmethyl phenol, alpha-phenyl-o-cresol, o-cresol, .alpha.-phenyl, unii-sf555bl24o, 2-benzyl phenol
PubChem CID 24216
IUPAC Name 2-benzylphenol
SMILES C1=CC=C(C=C1)CC2=CC=CC=C2O

Specifications

Melting Point 54°C
Boiling Point 312°C
Quantity 500 g
Formula Weight 184.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.