Learn More
Medchemexpress LLC HY-W044764 500mg Medchemexpress, 2-Benzylsuccinic acid CAS:884-33-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW044764500MG
Encompass_Preferred
Medchemexpress, HY-W044764 500mg 2-Benzylsuccinic acid CAS:884-33-3 2-Benzylsuccinic acid (DL-Benzylsuccinic acid) is an potent inhibitor of carboxypeptidase A (CPA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3483