Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Benzylsuccinic acid | 884-33-3 | MFCD00055798 | 99.8% | 208.21 g/mol | C11H12O4 | 1 ML
SDP

Catalog No. 5000210196
Encompass_Preferred

2-Benzylsuccinic acid is a small-molecule inhibitor of carboxypeptidase A and Nna1 supplied as a 10 mM solution in DMSO (1 mL). It is used in research on neuropathic pain (reduces cold hyperalgesia) and non-alcoholic steatohepatitis, and is provided at high purity for biochemical and pharmacological studies.

  • Provided as a ready-to-use 10 mM solution in DMSO.
  • High purity: 99.8%.
  • CAS number 884-33-3 for unambiguous identification.
  • Chemical formula C11H12O4 and molecular weight 208.21 g/mol.
  • Intended for research on carboxypeptidase inhibition and neuropathic pain models.

Catalog No. 50-002-10196 Supplier Medchemexpress LLC Supplier No. HYW04476410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Benzylsuccinic acid is a small-molecule inhibitor of carboxypeptidase A and Nna1 supplied as a 10 mM solution in DMSO (1 mL). It is used in research on neuropathic pain (reduces cold hyperalgesia) and non-alcoholic steatohepatitis, and is provided at high purity for biochemical and pharmacological studies.

  • Provided as a ready-to-use 10 mM solution in DMSO.
  • High purity: 99.8%.
  • CAS number 884-33-3 for unambiguous identification.
  • Chemical formula C11H12O4 and molecular weight 208.21 g/mol.
  • Intended for research on carboxypeptidase inhibition and neuropathic pain models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.