Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-benzylsuccinic acid (DL-benzylsuccinic acid) | 884-33-3 | 208.21 g/mol | C11H12O4 | 1 G
SDP

Catalog No. 5000210173
Encompass_Preferred

2-benzylsuccinic acid (DL-benzylsuccinic acid) is a research-grade small molecule reported as an orally active carboxypeptidase A and Nna1 inhibitor used in preclinical studies of neuropathic pain and metabolic disease. It is supplied as a solid and is also available as a 10 mM solution in DMSO.

  • Cas number: 884-33-3
  • Molecular formula: C11H12O4
  • Molecular weight: 208.21 g/mol
  • Available as solid quantities (including 1 g) and as DMSO solution stocks
  • Used in research on neuropathic pain (reduces cold hyperalgesia) and non-alcoholic steatohepatitis models

Catalog No. 50-002-10173 Supplier Medchemexpress LLC Supplier No. HYW0447641G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-benzylsuccinic acid (DL-benzylsuccinic acid) is a research-grade small molecule reported as an orally active carboxypeptidase A and Nna1 inhibitor used in preclinical studies of neuropathic pain and metabolic disease. It is supplied as a solid and is also available as a 10 mM solution in DMSO.

  • Cas number: 884-33-3
  • Molecular formula: C11H12O4
  • Molecular weight: 208.21 g/mol
  • Available as solid quantities (including 1 g) and as DMSO solution stocks
  • Used in research on neuropathic pain (reduces cold hyperalgesia) and non-alcoholic steatohepatitis models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.