Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Benzylthio)ethanol, 98%

Catalog No. AAL1227406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1227406 5 g
AAL1227414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1227406 Supplier Thermo Scientific Chemicals Supplier No. L1227406
Only null left
Add to Cart

CAS: 26524-88-9 | C9H12OS | 168.254 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26524-88-9
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00014039
InChI Key ANMHSIXPUAKNLM-UHFFFAOYSA-N
Synonym 2-benzylthio ethanol, 2-hydroxyethyl benzyl sulfide, 2-benzylsulfanyl ethanol, 2-hydroxyethyl-benzyl sulphide, 2-benzylsulfanyl ethan-1-ol, 2-phenylmethylthio ethanol, 2-phenylmethylsulfanyl ethanol, 2-phenylmethylthio ethan-1-ol, 2-hydroxyethylbenzylsulfide, 2-benzylthioethanol
PubChem CID 77486
IUPAC Name 2-benzylsulfanylethanol
SMILES C1=CC=C(C=C1)CSCCO

Specifications

Boiling Point 168°C to 169°C (18 mmHg)
Refractive Index 1.576
Linear Formula C6H5CH2S(CH2)2OH
Quantity 5 g
Beilstein 2356895
Formula Weight 168.25
Percent Purity 98%
Chemical Name or Material 2-(Benzylthio)ethanol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.