Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Benzylthio)ethanol 98.0+%, TCI America™
SDP

Catalog No. B40875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40875G 5 g
B408725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40875G Supplier TCI America Supplier No. B40875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3878-41-9
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00014039
InChI Key ANMHSIXPUAKNLM-UHFFFAOYSA-N
Synonym 2-Hydroxyethyl Benzyl Sulfide
PubChem CID 77486
IUPAC Name 2-benzylsulfanylethanol
SMILES C1=CC=C(C=C1)CSCCO

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 168.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Benzylthio)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.