Learn More
Medchemexpress LLC 2-Biphenylboronic acid | 4688-76-0 | 198.03 | 25 G
SDP
Catalog No. 5000449959
Encompass_Preferred
2-Biphenylboronic acid is a biochemical reagent that serves as a biological material or organic compound for life science related research.
  • Used as a substrate in palladium-catalyzed cross-coupling with benzylic acetates to provide diarylmethanes.
  • Acts as a reactant in Suzuki-Miyaura cross-coupling reactions with arylhalides, dibromovinyl precursors, alkenyl tosylates and mesylates, and quinoline carboxylates.
  • Involved in intramolecular Friedel-Crafts alkylation for the synthesis of chiral tetralins.
  • Utilized in hydroxylation to phenols and oxidative coupling with alkynes.
Catalog No. 50-004-49959 Supplier Medchemexpress LLC Supplier No. 25GHYW000939
2-Biphenylboronic acid is a biochemical reagent that serves as a biological material or organic compound for life science related research.
  • Used as a substrate in palladium-catalyzed cross-coupling with benzylic acetates to provide diarylmethanes.
  • Acts as a reactant in Suzuki-Miyaura cross-coupling reactions with arylhalides, dibromovinyl precursors, alkenyl tosylates and mesylates, and quinoline carboxylates.
  • Involved in intramolecular Friedel-Crafts alkylation for the synthesis of chiral tetralins.
  • Utilized in hydroxylation to phenols and oxidative coupling with alkynes.