Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Biphenylyl glycidyl ether, 95%, Thermo Scientific™

Catalog No. AC188172500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC188172500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC188172500 Supplier Thermo Scientific Chemicals Supplier No. 188172500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7144-65-2
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00046989
InChI Key DNVXWIINBUTFEP-UHFFFAOYNA-N
Synonym 2-biphenylyl glycidyl ether, opp-g, 3-2-xenyloxy propylene oxide, 2-1,1'-biphenyl-2-yloxy methyl oxirane, ccris 2065, 3-2-biphenyloxy propylene oxide, 3-2-xenolyl-1,2-epoxypropane, biphenyl-2-yl 2,3-epoxypropyl ether, ether, 2-biphenylyl 2,3-epoxypropyl, oxirane, 2-1,1'-biphenyl-2-yloxy methyl
PubChem CID 97875
IUPAC Name 2-({[1,1'-biphenyl]-2-yloxy}methyl)oxirane
SMILES C(OC1=CC=CC=C1C1=CC=CC=C1)C1CO1

Specifications

Melting Point 30.0°C to 32.0°C
Boiling Point 120.0°C (0.1 mmHg)
Quantity 250 g
Formula Weight 226.27
Chemical Name or Material 2-Biphenylyl glycidyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.