Learn More
Medchemexpress LLC 2-biphenylyl phenyl phosphate | 132-29-6 | MFCD01705737 | 98.0% | 402.38 | C24H19O4P | 100 MG
SDP
Catalog No. 5000280620
Encompass_Preferred
2-Biphenylyl phenyl phosphate is an aryl phosphate ester used primarily as a flame retardant and as an additive in plastics. It is supplied for research and formulation work where aromatic phosphate esters are required, and it is chosen for applications that need improved flame resistance and compatibility with polymer matrices.
  • Used as a flame retardant in plastics and polymers.
  • Functions as a plastic additive to modify processing and performance.
  • Provides thermal stability from its aromatic phosphate structure.
  • Compatible with common polymer matrices for formulation work.
  • Supplied in small research quantities suitable for testing and development.
Catalog No. 50-002-80620 Supplier Medchemexpress LLC Supplier No. HYW292950100MG
2-Biphenylyl phenyl phosphate is an aryl phosphate ester used primarily as a flame retardant and as an additive in plastics. It is supplied for research and formulation work where aromatic phosphate esters are required, and it is chosen for applications that need improved flame resistance and compatibility with polymer matrices.
  • Used as a flame retardant in plastics and polymers.
  • Functions as a plastic additive to modify processing and performance.
  • Provides thermal stability from its aromatic phosphate structure.
  • Compatible with common polymer matrices for formulation work.
  • Supplied in small research quantities suitable for testing and development.