Learn More
Medchemexpress LLC 2-biphenylyl phenyl phosphate | 132-29-6 | MFCD01705737 | 98.0% | 402.38 g/mol | C24H19O4P | 25 MG
SDP
Catalog No. 5000282528
Encompass_Preferred
2-Biphenylyl phenyl phosphate is an organophosphate ester used primarily as a flame retardant and plastic additive. It is incorporated into polymer formulations to impart fire resistance and modify material performance. CAS 132-29-6; formula C24H19O4P; molecular weight 402.38 g/mol.
  • Provides flame retardancy in polymer and plastic formulations.
  • Acts as a plastic additive to improve material performance.
  • Aromatic phosphate ester compatible with many thermoplastics.
  • Stable under typical processing conditions for additives.
  • Well-documented chemical identifiers for formulation work.
Catalog No. 50-002-82528 Supplier Medchemexpress LLC Supplier No. HYW29295025MG
2-Biphenylyl phenyl phosphate is an organophosphate ester used primarily as a flame retardant and plastic additive. It is incorporated into polymer formulations to impart fire resistance and modify material performance. CAS 132-29-6; formula C24H19O4P; molecular weight 402.38 g/mol.
  • Provides flame retardancy in polymer and plastic formulations.
  • Acts as a plastic additive to improve material performance.
  • Aromatic phosphate ester compatible with many thermoplastics.
  • Stable under typical processing conditions for additives.
  • Well-documented chemical identifiers for formulation work.